Anti-Protocadherin Fat 1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13165-100EA 341 EUR
BIRBORB13165-100
Anti-Protocadherin Fat 1 Rabbit Polyclonal Antibody
Anticuerpos
W1:2000-5000; Immunofluorescence: 1:100

Type: Primary
Antigen: Protocadherin Fat 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR