Anti-LPA receptor 1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13151-100EA 341 EUR
BIRBORB13151-100
Anti-LPA receptor 1 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:1000

Type: Primary
Antigen: LPA receptor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR