Anti-alpha 1 IL13 Receptor Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13150-100EA 0 EUR
BIRBORB13150-100
Anti-alpha 1 IL13 Receptor Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1000

Type: Primary
Antigen: alpha 1 IL13 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR