Anti-SPP1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13123-100EA 341 EUR
BIRBORB13123-100
Anti-SPP1 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:1000

Type: Primary
Antigen: SPP1 (secreted phosphoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR