Anti-TUBB3 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13113-100EA 0 EUR
BIRBORB13113-100
Anti-TUBB3 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:5000-10000

Type: Primary
Antigen: TUBB3 (tubulin, beta 3 class III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR