Anti-CD24 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13106-100EA 0 EUR
BIRBORB13106-100
Anti-CD24 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD24 (CD24 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR