Anti-ZFP42 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13095-100EA 0 EUR
BIRBORB13095-100
Anti-ZFP42 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:5000

Type: Primary
Antigen: ZFP42 (ZFP42 zinc finger protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR