Anti-CD8A Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13084-100EA 0 EUR
BIRBORB13084-100
Anti-CD8A Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500; Immunofluorescence: 1:250

Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR