Anti-His tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Biorbyt
BIRBORB13075-100EA 385 EUR
BIRBORB13075-100
Anti-His tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: His tag (His tag)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR