Anti-SLC6A3 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12998-100EA 363 EUR
BIRBORB12998-100
Anti-SLC6A3 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SLC6A3 (solute carrier family 6 (neurotransmitter transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR