Anti-SLC1A7 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12975-100EA 0 EUR
BIRBORB12975-100
Anti-SLC1A7 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SLC1A7 (solute carrier family 1 (glutamate transporter), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR