Anti-KAT5 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12969-1EA 145 EUR
BIRBORB12969-1
Anti-KAT5 Mouse Monoclonal Antibody
Anticuerpos
Western Blot: 1:1000

Type: Primary
Antigen: KAT5 (K(lysine) acetyltransferase 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR