Anti-APP Goat polyclonal antibody

Proveedor: Biorbyt
BIRBORB12385-100EA 363 EUR
BIRBORB12385-100
Anti-APP Goat polyclonal antibody
Anticuerpos
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR