Anti-TRIM6 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12384-100EA 290 EUR
BIRBORB12384-100
Anti-TRIM6 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TRIM6 (tripartite motif containing 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR