Anti-TIAM2 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12382-100EA 290 EUR
BIRBORB12382-100
Anti-TIAM2 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TIAM2 (T-cell lymphoma invasion and metastasis 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR