Anti-HMHA1 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12381-100EA 290 EUR
BIRBORB12381-100
Anti-HMHA1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: HMHA1 (histocompatibility (minor) HA-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR