Anti-DCHS1 Goat polyclonal antibody

Proveedor: Biorbyt
BIRBORB12379-100EA 290 EUR
BIRBORB12379-100
Anti-DCHS1 Goat polyclonal antibody
Anticuerpos
Type: Primary
Antigen: DCHS1 (dachsous cadherin-related 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR