Anti-TRPC6 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12355-100EA 290 EUR
BIRBORB12355-100
Anti-TRPC6 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TRPC6 (transient receptor potential cation channel, subfamily C, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR