Anti-ABHD14B Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12328-100EA 363 EUR
BIRBORB12328-100
Anti-ABHD14B Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ABHD14B (abhydrolase domain containing 14B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR