Anti-SYT9 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12323-100EA 363 EUR
BIRBORB12323-100
Anti-SYT9 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SYT9 (synaptotagmin IX)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR