Anti-TAS2R4 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12319-100EA 0 EUR
BIRBORB12319-100
Anti-TAS2R4 Goat Polyclonal Antibody
Anticuerpos
ELISA: 1:128000

Type: Primary
Antigen: TAS2R4 (taste receptor, type 2, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR