Anti-LSM2 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12304-100EA 0 EUR
BIRBORB12304-100
Anti-LSM2 Goat Polyclonal Antibody
Anticuerpos
ELISA: 1:2000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: LSM2 (LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR