Anti-ENC1 Goat polyclonal antibody

Proveedor: Biorbyt
BIRBORB12293-100EA 363 EUR
BIRBORB12293-100
Anti-ENC1 Goat polyclonal antibody
Anticuerpos
ELISA: 1:128000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: ENC1 (ectodermal-neural cortex 1 (with BTB domain))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR