Anti-CALB1 Goat polyclonal antibody

Proveedor: Biorbyt
BIRBORB12289-100EA 363 EUR
BIRBORB12289-100
Anti-CALB1 Goat polyclonal antibody
Anticuerpos
ELISA: 1:128000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: CALB1 (calbindin 1, 28kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR