Anti-LMNB1 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12278-100EA 363 EUR
BIRBORB12278-100
Anti-LMNB1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LMNB1 (lamin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR