Anti-FGF14 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12272-100EA 363 EUR
BIRBORB12272-100
Anti-FGF14 Goat Polyclonal Antibody
Anticuerpos
ELISA: 1:128000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: FGF14 (fibroblast growth factor 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR