Anti-RPL23 Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12263-100EA 363 EUR
BIRBORB12263-100
Anti-RPL23 Goat Polyclonal Antibody
Anticuerpos
ELISA: 1:128000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: RPL23 (ribosomal protein L23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR