Anti-V5 Tag E. coli Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12256-100EA 276 EUR
BIRBORB12256-100 BIRBORB12256-50
Anti-V5 Tag E. coli Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: V5 Tag (V5 tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: E. coli
Isotype: IgG1 (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR