Anti-HSV Tag E. coli Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12255-50EA 218 EUR
BIRBORB12255-50 BIRBORB12255-100
Anti-HSV Tag E. coli Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: HSV Tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: E. coli
Isotype: IgG1 (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR