Anti-HA tag E. coli Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12254-100EA 276 EUR
BIRBORB12254-100 BIRBORB12254-50
Anti-HA tag E. coli Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: E. coli
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR