Anti-IgG Goat Polyclonal Antibody (Biotin)

Proveedor: Biorbyt
BIRBORB12242-100EA 218 EUR
BIRBORB12242-100
Anti-IgG Goat Polyclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR