Anti-IgG Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB12224-100EA 218 EUR
BIRBORB12224-100
Anti-IgG Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR