Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Biorbyt
BIRBORB12222-100EA 218 EUR
BIRBORB12222-100
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR