Anti-IgG2b Mouse Monoclonal Antibody (APC (Allophycocyanin))

Proveedor: Biorbyt
BIRBORB12201-100EA 276 EUR
BIRBORB12201-100 BIRBORB12201-50
Anti-IgG2b Mouse Monoclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Type: Primary
Antigen: IgG2b
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR