Anti-IgG2b Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)

Proveedor: Biorbyt
BIRBORB12200-50EA 218 EUR
BIRBORB12200-50 BIRBORB12200-100
Anti-IgG2b Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Anticuerpos
Type: Primary
Antigen: IgG2b
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR