Anti-IgG2a Mouse Monoclonal Antibody (APC (Allophycocyanin))

Proveedor: Biorbyt
BIRBORB12196-100EA 276 EUR
BIRBORB12196-100 BIRBORB12196-50
Anti-IgG2a Mouse Monoclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Type: Primary
Antigen: IgG2a
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR