Anti-IgG1 Mouse Monoclonal Antibody (APC (Allophycocyanin))

Proveedor: Biorbyt
BIRBORB12189-50EA 218 EUR
BIRBORB12189-50 BIRBORB12189-100
Anti-IgG1 Mouse Monoclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Type: Primary
Antigen: IgG1
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR