Anti-CD8A Rat Monoclonal Antibody (APC (Allophycocyanin))

Proveedor: Biorbyt
BIRBORB12183-50EA 240 EUR
BIRBORB12183-50 BIRBORB12183-100
Anti-CD8A Rat Monoclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR