Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein))

Proveedor: Biorbyt
BIRBORB12180-100EA 312 EUR
BIRBORB12180-100 BIRBORB12180-50
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR