Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin))

Proveedor: Biorbyt
BIRBORB12176-50EA 240 EUR
BIRBORB12176-50 BIRBORB12176-100
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR