Anti-CD5/CD19 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)

Proveedor: Biorbyt
BIRBORB12164-20EA 218 EUR
BIRBORB12164-20 BIRBORB12164-50
Anti-CD5/CD19 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)
Anticuerpos
Type: Primary
Antigen: CD5/CD19
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1+IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR