Anti-CD40LG Mouse Monoclonal Antibody (PE (Phycoerythrin))
Proveedor: Biorbyt
BIRBORB12146-50EA 0 EUR
BIRBORB12146-50 BIRBORB12146-100
Anti-CD40LG Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Type: Primary
Antigen: CD40LG (CD40 ligand)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human