Anti-CTLA4 Mouse monoclonal antibody

Proveedor: Biorbyt
BIRBORB12141-100EA 0 EUR
BIRBORB12141-100 BIRBORB12141-500
Anti-CTLA4 Mouse monoclonal antibody
Anticuerpos
Type: Primary
Antigen: CTLA4 (cytotoxic T-lymphocyte-associated protein 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR