Anti-ENG Mouse monoclonal antibody

Proveedor: Biorbyt
BIRBORB12132-100EA 218 EUR
BIRBORB12132-100 BIRBORB12132-500
Anti-ENG Mouse monoclonal antibody
Anticuerpos
Type: Primary
Antigen: ENG (endoglin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR