Anti-CD58 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12100-100EA 218 EUR
BIRBORB12100-100 BIRBORB12100-500
Anti-CD58 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD58 (CD58 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR