Anti-CD55 Mouse Monoclonal Antibody (FITC (Fluorescein))

Proveedor: Biorbyt
BIRBORB12092-50EA 240 EUR
BIRBORB12092-50 BIRBORB12092-100
Anti-CD55 Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: CD55 (CD55 molecule, decay accelerating factor for complement (Cromer blood group))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR