Anti-ITGA4 Mouse Monoclonal Antibody (FITC (Fluorescein))

Proveedor: Biorbyt
BIRBORB12083-100EA 312 EUR
BIRBORB12083-100 BIRBORB12083-50
Anti-ITGA4 Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR