Anti-ITGA4 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12082-100EA 0 EUR
BIRBORB12082-100 BIRBORB12082-500
Anti-ITGA4 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR