Anti-PTPRC Mouse Monoclonal Antibody (PE (Phycoerythrin))

Proveedor: Biorbyt
BIRBORB12062-50EA 0 EUR
BIRBORB12062-50 BIRBORB12062-100
Anti-PTPRC Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR