Anti-CD44 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB12057-100EA 218 EUR
BIRBORB12057-100 BIRBORB12057-500
Anti-CD44 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR